Copper(I) thiocyanate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide

93-2964-250G 93-2964-50G
100202-062EA 225.05 USD
100202-062 100202-064
Copper(I) thiocyanate ≥99%
Copper(I) thiocyanate
Formula: CCuNS
MW: 121.63 g/mol
Melting Pt: 1084 °C
Density: 2.843
MDL Number: MFCD00010980
CAS Number: 1111-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR