Copper(II) tetrafluoroborate hydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Copper(II) tetrafluoroborate xhydrate

93-2963-250G 93-2963-50G
100200-118EA 165.6 USD
100200-118 100200-120
Copper(II) tetrafluoroborate hydrate ≥99%
Copper(II) tetrafluoroborate hydrate
Formula: B₂CuF₈H₂O
MW: 255.170 g/mol
MDL Number: MFCD00150223
CAS Number: 207121-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR