Copper(II) tartrate hydrate

Supplier: STREM CHEMICALS INC MS

93-2961-50G 93-2961-250G
100205-874EA 216.55 USD
100205-874 100200-332
Copper(II) tartrate hydrate
Copper(II) tartrate hydrate
Formula: C₄H₄CuO₆·xH₂O
MDL Number: MFCD00150650
CAS Number: 946843-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR