Copper(II) oxalate hemihydrate

Supplier: STREM CHEMICALS INC MS

Synonyms: Cupric oxalate hemihydrate

93-2942-100G 93-2942-25G
100199-546EA 159.94 USD
100199-546 100196-600
Copper(II) oxalate hemihydrate
Copper(II) oxalate hemihydrate
Formula: CuC₂O₄·½H₂O
MW: 160.57 g/mol (151.51 anhy)
MDL Number: MFCD00150451
CAS Number: 5893-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR