Copper(II) hexafluoroacetylacetonate hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hexafluoroacetylacetono Copper(II) Salt, Copper(II) Hexafluoroacetylacetonate, Copper(II) hexafluoro-2,4-pentanedionate hydrate, Bis(hexafluoroacetylacetonato)copper(II) hydrate, Copper(II) hexafluoro-2,4-pentanedionate xhydrate

93-2929-5G 93-2929-25G 93-2929-1G
100199-062EA 125.97 USD
100199-062 100199-060 100207-016
Copper(II) hexafluoroacetylacetonate hydrate
Copper (II) hexafluoroacetylacetonate hydrate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₀H₂CuF₁₂O₄·xH₂O
MW: 477.64 g/mol (anhy)
Boiling Pt: 220 °C/ 100 °C (0.5 mmHg) (dec./ subl.)
Melting Pt: 97…99 °C
MDL Number: MFCD00151019
CAS Number: 155640-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR