Copper(II) ethylacetoacetate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-2920-5G 93-2920-25G
100200-114EA 182.58 USD
100200-114 100200-112
Copper(II) ethylacetoacetate ≥99%
Copper(II) ethylacetoacetate
Formula: (CH₃CH₂CO₂CHCOCH₃)₂Cu
MW: 321.81 g/mol
MDL Number: MFCD00017759
CAS Number: 14284-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR