Ammonium tetrachlorocuprate dihydrate
Supplier: STREM CHEMICALS INC MS
Synonyms:
Ammonium copper(II) chloride dihydrate
93-2902-500G
93-2902-100G
100210-334EA
261.85
USD
100210-334
100196-590
Ammonium tetrachlorocuprate dihydrate
Ammonium tetrachlorocuprate dihydrate
Formula:
H₈Cl₄CuN₂·2H₂O MW: 277.46 Melting Pt: 110 °C Density: 1.993 |
MDL Number:
MFCD00150384 CAS Number: 10060-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...