Nickel boride ≥99% (Ni basis)
Supplier: STREM CHEMICALS INC MS
93-2859-50G
93-2859-10G
100203-436EA
348.18
USD
100203-436
100205-488
Nickel boride ≥99% (Ni basis)
Nickel boride
Formula:
BNi MW: 69.50 g/mol Density: 7,39 |
MDL Number:
MFCD00016261 CAS Number: 12007-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...