Nickel(II) hydroxide

Supplier: STREM CHEMICALS INC MS
Danger

93-2847-250G 93-2847-1000G
100211-248EA 157.11 USD
100211-248 100200-108
Nickel(II) hydroxide
Nickel(II) hydroxide
Formula: H₂NiO₂
MW: 92.71 g/mol
Melting Pt: 230 °C (dec.)
MDL Number: MFCD00011140
CAS Number: 12054-48-7
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR