Nickel(II) tetrafluoroborate hexahydrate ≥99%

Supplier: STREM CHEMICALS INC MS

93-2841-50G 93-2841-250G
100209-072EA 93.42 USD
100209-072 100209-070
Nickel(II) tetrafluoroborate hexahydrate ≥99%
Nickel(II) tetrafluoroborate hexahydrate
Formula: B₂F₈Ni·6H₂O
MW: 340.394 g/mol
Density: 1.47
MDL Number: MFCD00150259
CAS Number: 15684-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR