Potassium tetracyanonickelate(II) hydrate

Supplier: STREM CHEMICALS INC MS
Danger

93-2836-50G 93-2836-10G
100203-848EA 561.91 USD
100203-848 100208-146
Potassium tetracyanonickelate(II) hydrate
Potassium tetracyanonickelate(II) hydrate
Formula: K₂[Ni(CN)₄].xH₂O
MW: 240.96 g/mol (anhy)
MDL Number: MFCD00049669
CAS Number: 339527-86-5
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR