Nickel(II) 2-ethylhexanoate ≥78% in 2-ethylhexanoic acid (10-15% Ni)

Supplier: STREM CHEMICALS INC MS

93-2830-250G 93-2830-50G
100209-066EA 1432.37 USD
100209-066 100197-794
Nickel(II) 2-ethylhexanoate ≥78% in 2-ethylhexanoic acid (10-15% Ni)
Nickel(II) 2-ethylhexanoate
Density: 1.08
Flash Pt: 155 °F
MDL Number: MFCD00014002
CAS Number: 4454-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR