Nickel naphthenate ∼60% in toluene (6-8% Ni)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Nickel(II) naphthenate
93-2829-250G
93-2829-50G
100199-050EA
476.98
USD
100199-050
100199-052
Nickel naphthenate ∼60% in toluene (6-8% Ni)
Nickel naphthenate
Formula:
C₂₂H₁₄NiO₄ MW: 401.043 g/mol |
MDL Number:
MFCD00147723 CAS Number: 61788-71-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...