Nickel(II) hexafluoroacetylacetonate hydrate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Hexafluoroacetylacetono Nickel(II) Salt, Bis(hexafluoroacetylacetonato)nickel(II) hydrate

93-2821-5G 93-2821-1G
100206-990EA 314.22 USD
100206-990 100205-480
Nickel(II) hexafluoroacetylacetonate hydrate ≥98%
Nickel(II) hexafluoroacetylacetonate hydrate
Formula: C₁₀H₂F₁₂NiO₄·xH₂O
MW: 472.80 g/mol (anhy)
Melting Pt: 211…213 °C
MDL Number: MFCD00067506
CAS Number: 14949-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR