Nickel dimethylglyoxime ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Nickel(II) dimethylglyoxime

93-2815-10G 93-2815-50G
100204-278EA 93.42 USD
100204-278 100202-040
Nickel dimethylglyoxime ≥99%
Nickel dimethylglyoxime
Formula: C₈H₁₄N₄NiO₄
MW: 288.915 g/mol
Melting Pt: 250 °C (subl.)
MDL Number: MFCD00136028
CAS Number: 13478-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR