Nickel carbonate, basic hydrate

Supplier: STREM CHEMICALS INC MS

93-2808-1000G 93-2808-250G 93-2808-50G
100198-350EA 369.42 USD
100198-350 100200-104 100210-322
Nickel carbonate, basic hydrate
Nickel carbonate, basic hydrate
Formula: NiCO₃·2Ni(OH)₂·xH₂O
MW: 304.12 g/mol (anhy)
MDL Number: MFCD00150262
CAS Number: 958638-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR