Nickel(II) chloride, dimethoxyethane adduct ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel(II) chloride ethylene glycol dimethyl ether complex

93-2801-25G 93-2801-5G
100211-238EA 557.66 USD
100211-238 100211-240
Nickel(II) chloride, dimethoxyethane adduct ≥97%
Nickel(II) chloride, dimethoxyethane adduct
Formula: C₄H₁₀Cl₂NiO₂
MW: 219.7204 g/mol
MDL Number: MFCD00013481
CAS Number: 29046-78-4
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR