Cobalt(II) wolframate ≥99.9% (Co basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Cobalt(II) tungsten oxide, Cobalt(II) tungstate

93-2754-10G 93-2754-50G
100204-272EA 106.15 USD
100204-272 100199-044
Cobalt(II) wolframate ≥99.9% (Co basis)
Cobalt(II) wolframate
Formula: CoO₄W
MW: 306.769 g/mol
Density: 8.42
MDL Number: MFCD00016034
CAS Number: 10101-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR