Cobalt(II) stearate (9 - 10% Co)

Supplier: STREM CHEMICALS INC MS

Synonyms: Cobalt(II) Stearate, Cobalt stearate

93-2714-500G 93-2714-2000G
100200-744EA 117.48 USD
100200-744 100200-742
Cobalt(II) stearate (9 - 10% Co)
Cobalt(II) stearate
Formula: C₃₆H₇₀CoO₄
MW: 625.872 g/mol
MDL Number: MFCD00054343
CAS Number: 1002-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR