Cobalt(II) citrate hydrate

Supplier: STREM CHEMICALS INC MS

93-2707-1000G 93-2707-250G
100200-094EA 928.49 USD
100200-094 100202-546
Cobalt(II) citrate hydrate
Cobalt(II) citrate hydrate
MDL Number: MFCD00150521
CAS Number: 866-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR