Iron(III) perchlorate hydrate, purple

Supplier: STREM CHEMICALS INC MS
Danger

93-2673-50G 93-2673-250G
100207-818EA 45.29 USD
100207-818 100205-460
Iron(III) perchlorate hydrate, purple
Iron(III) perchlorate hydrate
Formula: Cl₃FeH₂O₁₃
MW: 372.198 g/mol
MDL Number: MFCD00149705
CAS Number: 15201-61-3
UN: 1481
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR