Iron(III) fluoride trihydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-2670-100G 93-2670-25G
100209-424EA 203.82 USD
100209-424 100197-778
Iron(III) fluoride trihydrate ≥98%
Iron(III) fluoride trihydrate
Formula: F₃Fe·3H₂O
MW: 166.89 g/mol
MDL Number: MFCD00149718
CAS Number: 15469-38-2
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR