Iron(III) 1,1,1-trifluoro-2,4-pentanedionate ≥99% (99.9%-Fe)

Supplier: STREM CHEMICALS INC MS
Warning

93-2644-5G 93-2644-25G
100210-298EA 217.97 USD
100210-298 100204-690
Iron(III) 1,1,1-trifluoro-2,4-pentanedionate ≥99% (99.9%-Fe)
Iron(III) 1,1,1-trifluoro-2,4-pentanedionate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₅H₁₂F₉FeO₆
Melting Pt: 110…112 °C
MDL Number: MFCD00017760
CAS Number: 14526-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR