Iron(II) sulfamate (38 - 42%) in aqueous solution

Supplier: STREM CHEMICALS INC MS

93-2638-1000G 93-2638-250G
100208-708EA 308.55 USD
100208-708 100196-572
Iron(II) sulfamate (38 - 42%) in aqueous solution
Iron(II) sulfamate
MDL Number: MFCD00055536
CAS Number: 14017-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR