Iron(II) tetrafluoroborate 40 - 45% in aqueous solution

Supplier: STREM CHEMICALS INC MS
Warning

93-2633-50G 93-2633-250G
100196-570EA 67.94 USD
100196-570 100210-296
Iron(II) tetrafluoroborate 40 - 45% in aqueous solution
Iron(II) tetrafluoroborate
MDL Number: MFCD00016079
CAS Number: 15283-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR