Iron(III) oxide monohydrate, yellow ≥99.9% (Fe basis)

Supplier: STREM CHEMICALS INC MS

93-2618-5000G 93-2618-1000G
100199-534EA 161.35 USD
100199-534 100207-810
Iron(III) oxide monohydrate, yellow ≥99.9% (Fe basis)
Iron(III) oxide monohydrate
MDL Number: MFCD00149717
CAS Number: 51274-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR