Iron(III) oxalate hexahydrate, Technical Grade

Supplier: STREM CHEMICALS INC MS
Warning

93-2615-5G 93-2615-25G
100205-458EA 76.43 USD
100205-458 100209-054
Iron(III) oxalate hexahydrate, Technical Grade
Iron(III) oxalate hexahydrate
Formula: Fe₂(C₂O₄)₃·6H₂O
MDL Number: MFCD00054430
CAS Number: 166897-40-1
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR