Manganese(II) 2-ethylhexanoate 40% in mineral spirits (6% Mn)

Supplier: STREM CHEMICALS INC MS
Warning

93-2532-1000G 93-2532-250G
100210-282EA 181.17 USD
100210-282 100205-452
Manganese(II) 2-ethylhexanoate 40% in mineral spirits (6% Mn)
Manganese(II) 2-ethylhexanoate
Formula: C₁₆H₃₀MnO₄
MDL Number: MFCD00058698
CAS Number: 13434-24-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR