Manganese(II) sulfide ≥99.9% (Mn basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-2524-25G 93-2524-5G
100200-078EA 196.74 USD
100200-078 100200-080
Manganese(II) sulfide ≥99.9% (Mn basis)
Manganese (II) sulphide
Formula: MnS
MW: 87.00 g/mol
Melting Pt: Dec.
Density: 3,99
MDL Number: MFCD00014210
CAS Number: 18820-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR