Manganese(II) perchlorate hexahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-2520-500G 93-2520-100G
100209-418EA 261.85 USD
100209-418 100204-250
Manganese(II) perchlorate hexahydrate ≥99%
Manganese (II) perchlorate hexahydrate
Formula: Cl₂MnO₈·6H₂O
MDL Number: MFCD00150255
CAS Number: 15364-94-0
UN: 1481
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR