Manganese naphthenate 56% in mineral spirits (6% Mn)

Supplier: STREM CHEMICALS INC MS
Warning

93-2516-500G 93-2516-100G
100210-280EA 121.72 USD
100210-280 100205-448
Manganese naphthenate 56% in mineral spirits (6% Mn)
Manganese naphthenate
Formula: ₂(C₁₁H₇O₂).Mn
Flash Pt: 104 °F
MDL Number: MFCD00147533
CAS Number: 1336-93-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR