Manganese(II) cyclohexanebutyrate

Supplier: STREM CHEMICALS INC MS

93-2508-5G 93-2508-1G
100199-530EA 242.03 USD
100199-530 100199-024
Manganese(II) cyclohexanebutyrate
Manganese (II) cyclohexanebutyrate
Formula: C₂₀H₃₄MnO₄
MDL Number: MFCD00036401
CAS Number: 35542-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR