Trichromium dicarbide ≥99.7%

Supplier: STREM CHEMICALS INC MS

Synonyms: Chromium carbide

93-2432-50G 93-2432-250G
100202-000EA 66.52 USD
100202-000 100206-016
Trichromium dicarbide ≥99.7%
Trichromium dicarbide
Formula: C₂Cr₃
MW: 180.01 g/mol
Boiling Pt: 3800 °C
Melting Pt: 1890 °C
Density: 6.68
MDL Number: MFCD00016041
CAS Number: 12012-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR