Titanium diboride ≥99%

Supplier: STREM CHEMICALS INC MS

93-2242-25G 93-2242-100G
100201-976EA 49.54 USD
100201-976 100209-414
Titanium diboride ≥99%
Titanium diboride
Formula: B₂Ti
MW: 69.5 g/mol
Melting Pt: 2900 °C
Density: 4,50
MDL Number: MFCD00049595
CAS Number: 12045-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR