Dihydrogen hexafluorotitanate 60% in aqueous solution

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Hexafluorotitanic acid, Hydrogen hexafluorotitanate, Fluorotitanic acid

93-2230-500G 93-2230-2000G
100209-046EA 148.62 USD
100209-046 100196-546
Dihydrogen hexafluorotitanate 60% in aqueous solution
Dihydrogen hexafluorotitanate
Formula: H₂TiF₆
MW: 163.91 g/mol
MDL Number: MFCD00049647
CAS Number: 17439-11-1
UN: 3264
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR