Scandium(III) carbonate hydrate ≥99.99% (Sc basis), PURATREM

Supplier: STREM CHEMICALS INC MS

93-2110-1G 93-2110-5G
100197-738EA 287.32 USD
100197-738 100198-046
Scandium(III) carbonate hydrate ≥99.99% (Sc basis), PURATREM
Scandium(III) carbonate hydrate
Formula: Sc₂(CO₃)₃·xH₂O
MW: 269.94 g/mol (anhy)
MDL Number: MFCD00149955
CAS Number: 5809-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR