Calcium perchlorate hydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-2033-100G 93-2033-500G
100198-998EA 48.12 USD
100198-998 100203-842
Calcium perchlorate hydrate ≥99%
Calcium perchlorate hydrate
Formula: Ca(ClO₄)₂·xH₂O
MW: 238.98 g/mol (anhy)
MDL Number: MFCD00015971
CAS Number: 13477-36-6
UN: 1455
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR