Calcium iodate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-2023-250G 93-2023-50G
100201-960EA 140.12 USD
100201-960 100207-794
Calcium iodate ≥98%
Calcium iodate
Formula: CaI₂O₆
MW: 389.881 g/mol
Melting Pt: 540 °C (dec.)
Density: 4,519
MDL Number: MFCD00036265
CAS Number: 7789-80-2
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR