Tricalcium diarsenate 95

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Calcium arsenate

93-2004-500G 93-2004-25G 93-2004-100G
100204-214EA 1321.97 USD
100204-214 100200-532 100204-212
Tricalcium diarsenate 95
Tricalcium diarsenate
Formula: As₂Ca₃O₈
MW: 398.072 g/mol
Density: 3.620
MDL Number: MFCD00049407
CAS Number: 7778-44-1
UN: 1573
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR