Calcium acetylacetonate hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(2,4-pentanedionato)calcium(II), Acetylacetone Calcium(II) Salt, Calcium(II) Acetylacetonate, Calciumacetylacetonate

93-2002-25G 93-2002-100G
100196-534EA 66.52 USD
100196-534 100210-236
Calcium acetylacetonate hydrate
Calcium acetylacetonate hydrate
Formula: C₁₀H₁₄CaO₄·xH₂O
MW: 238.30 g/mol (anhy)
Melting Pt: 175 °C (dec.)
MDL Number: MFCD00150163
CAS Number: 19372-44-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR