Difluorophosphoric acid hemihydrate, Technical Grade

Supplier: STREM CHEMICALS INC MS
Danger

93-1571-1000G 93-1571-250G
100200-058EA 693.54 USD
100200-058 100211-222
Difluorophosphoric acid hemihydrate, Technical Grade
Difluorophosphoric acid hemihydrate
Boiling Pt: 115.9 °C
Melting Pt: –96.5 °C
Density: 1.583 (25°)
MDL Number: MFCD09842893
CAS Number: 13779-41-4
UN: 3264
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR