Triamyl phosphate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

93-1556-100G 93-1556-25G
100210-216EA 1222.89 USD
100210-216 100205-372
Triamyl phosphate ≥97%
Triamyl phosphate
Formula: C₁₅H₃₃O₄P
MDL Number: MFCD00015348
CAS Number: 2528-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR