Phenylphosphonic acid ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Phenylphosphonic Acid
93-1537-1000G
93-1537-250G
93-1537-50G
100201-930EA
993.6
USD
100201-930
100201-932
100210-212
Phenylphosphonic acid ≥98%
Phenylphosphonic acid
Formula:
C₆H₇O₃P MW: 158.09 g/mol Melting Pt: 163…166 °C |
MDL Number:
MFCD00002136 CAS Number: 1571-33-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...