Phenylphosphinic acid ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-1536-500G 93-1536-100G
100208-122EA 172.68 USD
100208-122 100205-368
Phenylphosphinic acid ≥99%
Phenylphosphinic acid
Formula: C₆H₇O₂P
MW: 142.092 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD00002131
CAS Number: 1779-48-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR