Diisopropyl phosphonate ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Diisopropyl phosphite, Di-i-propylphosphite, Diisopropyl hydrogen phosphite
93-1527-50G
93-1527-250G
100196-518EA
104.74
USD
100196-518
100210-210
Diisopropyl phosphonate ≥98%
Diisopropyl phosphonate
Formula:
C₆H₁₅O₃P MW: 166.16 g/mol Boiling Pt: 70…71 °C (9 mmHg) Density: 0,997 Flash Pt: 156 °F |
MDL Number:
MFCD00117905 CAS Number: 1809-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...