Dibutyl phosphonate ≥96%

Supplier: STREM CHEMICALS INC MS

Synonyms: Di-n-butylphosphite, Dibutyl phosphite

93-1518-50G 93-1518-250G
100197-714EA 50.95 USD
100197-714 100204-188
Dibutyl phosphonate ≥96%
Dibutyl phosphonate
Formula: C₈H₁₉O₃P
Boiling Pt: 118…119 °C (11 mmHg)
Density: 0,995
Flash Pt: 250 °F
MDL Number: MFCD00066633
CAS Number: 1809-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR