Allyltriphenylphosphonium bromide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-1501-25G 93-1501-100G
100196-512EA 56.62 USD
100196-512 100205-364
Allyltriphenylphosphonium bromide ≥99%
Allyltriphenylphosphonium bromide
Formula: C₂₁H₂₀BrP
MW: 383.261 g/mol
Melting Pt: 222…225 °C
MDL Number: MFCD00011808
CAS Number: 1560-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR