1,3-Divinyltetramethyldisiloxane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

93-1457-100G 93-1457-25G
100200-052EA 307.14 USD
100200-052 100211-216
1,3-Divinyltetramethyldisiloxane ≥97%
1,3-Divinyltetramethyldisiloxane
Formula: C₈H₁₈OSi₂
Boiling Pt: 139…140 °C
Density: 0,809
Flash Pt: 76 °F
MDL Number: MFCD00014933
CAS Number: 2627-95-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR