Silicon tetraacetate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1443-100G
93-1443-25G
100201-260EA
292.98
USD
100201-260
100206-940
Silicon tetraacetate ≥95%
Silicon tetraacetate
Formula:
C₈H₁₂O₈Si Boiling Pt: 148 °C (6 mmHg)/ 110 °C (dec./ subl.) Melting Pt: 160 °C (dec.) |
MDL Number:
MFCD00026184 CAS Number: 562-90-3 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...