Silica gel 70-200 mesh for TLC

Supplier: STREM CHEMICALS INC MS

93-1427-2000G 93-1427-500G
100208-107EA 397.72 USD
100208-107 100208-109
Silica gel 70-200 mesh for TLC
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR